2,6-Dibromo-4-chlorophenol 98%

Supplier: Ambeed
Warning

A457813-10G A457813-1G A457813-5G
77252-888EA 35 USD
77252-888 77252-890 77252-892
2,6-Dibromo-4-chlorophenol 98%
2,6-Dibromo-4-chlorophenol

Precaución: for Research Use Only

Formula: C₆H₃Br₂ClO
MDL Number: MFCD00029751
CAS Number: 5324-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR