3,3'-Dimethylbiphenyl 98%

Supplier: Ambeed
Warning

A457786-1G A457786-25G A457786-5G
77279-240EA 10 USD
77279-240 77279-242 77279-244
3,3'-Dimethylbiphenyl 98%
3,3'-Dimethylbiphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₄
MDL Number: MFCD00008534
CAS Number: 612-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR