(E)-1-Methoxy-4-(2-nitrovinyl)benzene 97%

Supplier: Ambeed
Warning

Synonyms: (E)-1-(4-Methoxyphenyl)-2-nitroethene

A457766-100G A457766-1G A457766-25G A457766-5G
77265-192EA 420.24 USD
77265-192 77265-194 77265-196 77265-198
(E)-1-Methoxy-4-(2-nitrovinyl)benzene 97%
trans-4-Methoxy-β-nitrostyrene

Precaución: for Research Use Only

Formula: C₉H₉NO₃
MW: 179.173 g/mol
MDL Number: MFCD00024826
CAS Number: 5576-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR