2-Pyrimidineacetic acid 95%

Supplier: Ambeed
Warning

A457571-1G A457571-250MG
77279-234EA 66 USD
77279-234 77279-236
2-Pyrimidineacetic acid 95%
2-Pyrimidineacetic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MDL Number: MFCD04973302
CAS Number: 66621-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR