3-Bromo-2-iodopyridine 98%

Supplier: Ambeed
Warning

A457554-250MG A457554-25G A457554-5G A457554-1G
77279-228EA 10 USD
77279-228 77279-230 77279-232 77748-927
3-Bromo-2-iodopyridine 98%
3-Bromo-2-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₃BrIN
MDL Number: MFCD08543471
CAS Number: 408502-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR