(S)-3,3'-Bis([1,1'-biphenyl]-4-yl)-1,1'-binaphthalene]-2,2'-diol ≥98%, ee 99%

Supplier: Ambeed
Warning

A457063-100MG A457063-1G A457063-250MG
77252-882EA 59.74 USD
77252-882 77252-884 77252-886
(S)-3,3'-Bis([1,1'-biphenyl]-4-yl)-1,1'-binaphthalene]-2,2'-diol ≥98%, ee 99%
(S)-3,3'-Bis([1,1'-biphenyl]-4-yl)-1,1'-binaphthalene]-2,2'-diol

Precaución: for Research Use Only

Formula: C₄₄H₃₀O₂
MW: 590.70 g/mol
CAS Number: 220486-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR