Methyl 2,3-difluoro-6-methylbenzoate 97%

Supplier: Ambeed
Warning

A456714-100MG A456714-1G A456714-250MG
77265-166EA 50 USD
77265-166 77265-168 77265-170
Methyl 2,3-difluoro-6-methylbenzoate 97%
Methyl 2,3-difluoro-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MDL Number: MFCD16250150
CAS Number: 1379107-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR