(R)-1-Benzyl 4-tert-butyl 2-(hydroxymethyl)piperazine-1,4-dicarboxylate 97%

Supplier: Ambeed
Warning

A456604-100MG A456604-1G A456604-5G
77241-084EA 30.9 USD
77241-084 77241-086 77241-088
(R)-1-Benzyl 4-tert-butyl 2-(hydroxymethyl)piperazine-1,4-dicarboxylate 97%
(R)-1-Benzyl 4-tert-butyl 2-(hydroxymethyl)piperazine-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₆N₂O₅
MW: 350.40 g/mol
MDL Number: MFCD09261339
CAS Number: 1217813-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR