2-Methoxy-6-methylbenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A456552-100MG
A456552-1G
A456552-250MG
77265-160EA
28.84
USD
77265-160
77265-162
77265-164
2-Methoxy-6-methylbenzaldehyde 97%
2-Methoxy-6-methylbenzaldehyde
Precaución: for Research Use Only
Formula:
C₉H₁₀O₂ MW: 150.174 g/mol |
MDL Number:
MFCD08235006 CAS Number: 54884-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...