Benzyl (4-hydroxybutyl)carbamate 98%

Supplier: Ambeed
Warning

A456419-1G A456419-250MG A456419-25G A456419-5G
77291-748EA 41 USD
77291-748 77291-750 77291-752 77291-754
Benzyl (4-hydroxybutyl)carbamate 98%
Benzyl (4-hydroxybutyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₃
MW: 223.26 g/mol
MDL Number: MFCD01861335
CAS Number: 17996-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR