2-(2-Aminoethyl)isoindoline-1,3-dione hydrochloride 97%

Supplier: Ambeed
Warning

A456416-10G A456416-250MG A456416-5G A456416-1G
77241-078EA 345.05 USD
77241-078 77241-080 77241-082 77572-440
2-(2-Aminoethyl)isoindoline-1,3-dione hydrochloride 97%
2-(2-Aminoethyl)isoindoline-1,3-dione hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂O₂
MW: 226.65 g/mol
MDL Number: MFCD08272836
CAS Number: 30250-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR