3-(3,4-Dihydroxyphenyl)propionic acid 95%

Supplier: Ambeed
Warning

Synonyms: Dihydrocaffeic acid

A456071-100G A456071-1G A456071-25G A456071-5G
77252-874EA 362.56 USD
77252-874 77252-876 77252-878 77252-880
3-(3,4-Dihydroxyphenyl)propionic acid 95%
3-(3,4-Dihydroxyphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
MDL Number: MFCD00002776
CAS Number: 1078-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR