2-(Morpholinomethyl)phenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine

A455910-1G A455910-250MG A455910-5G
77252-868EA 74.16 USD
77252-868 77252-870 77252-872
2-(Morpholinomethyl)phenylboronic acid pinacol ester 98%
2-(Morpholinomethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₆BNO₃
MW: 303.2043 g/mol
MDL Number: MFCD07781208
CAS Number: 876316-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR