NOD-IN-1 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 1-tosyl-1H-indole-2-carboxylate

A455827-10MG A455827-1MG A455827-25MG A455827-50MG A455827-5MG
77265-150EA 273.98 USD
77265-150 77265-152 77265-154 77265-156 77265-158
NOD-IN-1 97%
NOD-IN-1

Precaución: for Research Use Only

Formula: C₁₈H₁₇NO₄S
MW: 343.39 g/mol
MDL Number: MFCD30496705
CAS Number: 132819-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR