4-Fluoro-6-methoxypyrimidin-2-amine 95%

Supplier: Ambeed
Danger

A455210-100MG A455210-250MG
77291-726EA 48.41 USD
77291-726 77291-728
4-Fluoro-6-methoxypyrimidin-2-amine 95%
4-Fluoro-6-methoxypyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₅H₆FN₃O
MW: 143.12 g/mol
MDL Number: MFCD00233534
CAS Number: 130687-25-1
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR