(+)-Di-p-anisoyl-D-tartaric acid 98%

Supplier: Ambeed
Warning

Synonyms: (+)-Bis(4-methoxybenzoyl)-D-tartaric Acid, (2S,3S)-2,3-Bis((4-methoxybenzoyl)oxy)succinic acid

A454995-100G A454995-10G A454995-25G
77291-720EA 51.01 USD
77291-720 77291-722 77291-724
(+)-Di-p-anisoyl-D-tartaric acid 98%
(+)-Di-p-anisoyl-D-tartaric acid

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₁₀
MDL Number: MFCD07368366
CAS Number: 191605-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR