4',4''',4'''''-(1,3,5-Triazine-2,4,6-triyl)tris(([1,1'-biphenyl]-4-carboxylic acid)) 98%

Supplier: Ambeed
Warning

A454941-100MG A454941-1G A454941-250MG A454941-5G
77265-140EA 22.66 USD
77265-140 77265-142 77265-144 77616-966
4',4''',4'''''-(1,3,5-Triazine-2,4,6-triyl)tris(([1,1'-biphenyl]-4-carboxylic acid)) 98%
4',4''',4'''''-(1,3,5-Triazine-2,4,6-triyl)tris(([1,1'-biphenyl]-4-carboxylic acid))

Precaución: for Research Use Only

Formula: C₄₂H₂₇N₃O₆
MW: 669.68 g/mol
MDL Number: MFCD32667112
CAS Number: 1331756-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR