Methyl 2-methoxy-6-oxo-5,6,7,8-tetrahydroquinoline-5-carboxylate 97%

Supplier: Ambeed
Warning

A454839-100MG A454839-1G A454839-250MG A454839-5G
77265-132EA 26.78 USD
77265-132 77265-134 77265-136 77265-138
Methyl 2-methoxy-6-oxo-5,6,7,8-tetrahydroquinoline-5-carboxylate 97%
Methyl 2-methoxy-6-oxo-5,6,7,8-tetrahydroquinoline-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₄
MW: 235.23 g/mol
MDL Number: MFCD26397315
CAS Number: 130518-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR