Triallyl(methyl)silane 98%
Supplier: Ambeed
|
Danger
|
A454741-10G
A454741-1G
A454741-250MG
A454741-5G
77252-844EA
106.09
USD
77252-844
77252-846
77252-848
77252-850
Triallyl(methyl)silane 98%
Triallyl(methyl)silane
Precaución: for Research Use Only
Formula:
C₁₀H₁₈Si MW: 166.34 g/mol |
MDL Number:
MFCD00800604 CAS Number: 1112-91-0 UN: 1993 ADR: 3,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...