(2,6-Dichlorophenyl)acetamide 97%

Supplier: Ambeed
Warning

A454499-1G A454499-250MG A454499-5G
77252-830EA 22 USD
77252-830 77252-832 77252-834
(2,6-Dichlorophenyl)acetamide 97%
(2,6-Dichlorophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO
MDL Number: MFCD00082745
CAS Number: 78433-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR