1,2-Bis((2R,5R)-2,5-diethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate 98%

Supplier: Ambeed
Warning

A454400-100MG A454400-1G A454400-250MG
77252-824EA 189.52 USD
77252-824 77252-826 77252-828
1,2-Bis((2R,5R)-2,5-diethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate 98%
1,2-Bis((2R,5R)-2,5-diethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₆H₄₆BF₄P₂Rh
MW: 610.30 g/mol
MDL Number: MFCD17018828
CAS Number: 213343-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR