(2R,5R)-(+)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone ≥97%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (2R,5R)-5-Benzyl-2-(tert-butyl)-3-methylimidazolidin-4-one, (2R,5R)-(+)-5-Benzyl-2-tert-butyl-3-methyl-4-imidazolidinone

A454241-100MG A454241-1G A454241-250MG A454241-5G
77252-816EA 28.84 USD
77252-816 77252-818 77252-820 77252-822
(2R,5R)-(+)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone ≥97%, ee 99%
(2R,5R)-(+)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₂O
MW: 246.35 g/mol
MDL Number: MFCD08276842
CAS Number: 390766-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR