4-(1H-Imidazol-4-yl)aniline 95%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Aminophenyl)-1H-imidazole

A454095-100MG A454095-1G A454095-250MG
77291-708EA 42.23 USD
77291-708 77291-710 77291-712
4-(1H-Imidazol-4-yl)aniline 95%
4-(1H-Imidazol-4-yl)aniline

Precaución: for Research Use Only

Formula: C₉H₉N₃
MW: 159.188 g/mol
MDL Number: MFCD00297065
CAS Number: 29528-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR