5-Benzyloxyindole-3-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-(Benzyloxy)-1H-indole-3-carbaldehyde, 5-benzyloxy-1h-indole-3-carbaldehyde, 5-Benzyloxyindole-3-carboxaldehyde

A453918-25G A453918-5G
77279-210EA 110.21 USD
77279-210 77279-212
5-Benzyloxyindole-3-carbaldehyde 98%
5-Benzyloxyindole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₃NO₂
MW: 251.285 g/mol
MDL Number: MFCD00014562
CAS Number: 6953-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR