5-Methylisatoic anhydride 96%

Supplier: Ambeed
Warning

Synonyms: 6-Methyl-1H-benzo[d][1,3]oxazine-2,4-dione

A452807-10G A452807-1G A452807-250MG A452807-25G A452807-5G
77265-112EA 135 USD
77265-112 77265-114 77265-116 77265-118 77265-120
5-Methylisatoic anhydride 96%
5-Methylisatoic anhydride

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.1568 g/mol
MDL Number: MFCD06809783
CAS Number: 4692-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR