2-(Bromomethyl)-2-phenyl-1,3-dioxolane 95%

Supplier: Ambeed
Warning

A452585-100MG A452585-1G A452585-250MG
77241-056EA 23 USD
77241-056 77241-058 77241-060
2-(Bromomethyl)-2-phenyl-1,3-dioxolane 95%
2-(Bromomethyl)-2-phenyl-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD00225209
CAS Number: 3418-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR