3-Iodo-5-methoxy-1H-indole, N-BOC protected 98%

Supplier: Ambeed
Warning

A452381-100MG A452381-1G A452381-250MG
77279-196EA 5 USD
77279-196 77279-198 77279-200
3-Iodo-5-methoxy-1H-indole, N-BOC protected 98%
3-Iodo-5-methoxy-1H-indole, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₄H₁₆INO₃
MDL Number: MFCD05864763
CAS Number: 192189-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR