(R)-2-((6R,8R)-7,7-Dimethyl-5,6,7,8-tetrahydro-6,8-methanoquinolin-2-yl)-4-phenyl-4,5-dihydrooxazole 98%

Supplier: Ambeed
Warning

A452306-100MG A452306-250MG A452306-50MG A452306-1G
77265-106EA 113.3 USD
77265-106 77265-108 77265-110 77748-917
(R)-2-((6R,8R)-7,7-Dimethyl-5,6,7,8-tetrahydro-6,8-methanoquinolin-2-yl)-4-phenyl-4,5-dihydrooxazole 98%
(R)-2-((6R,8R)-7,7-Dimethyl-5,6,7,8-tetrahydro-6,8-methanoquinolin-2-yl)-4-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₁H₂₂N₂O
MW: 318.41 g/mol
CAS Number: 1027754-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR