1-tert-Butyl-4-piperidone 98%

Supplier: Ambeed
Warning

A452214-10G A452214-250MG
77241-052EA 175 USD
77241-052 77241-054
1-tert-Butyl-4-piperidone 98%
1-tert-Butyl-4-piperidone

Precaución: for Research Use Only

Formula: C₉H₁₇NO
MDL Number: MFCD03411608
CAS Number: 1465-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR