2-(Thiophen-2-yl)quinoxaline 97%

Supplier: Ambeed
Warning

A452071-1G A452071-250MG
77241-048EA 361.53 USD
77241-048 77241-050
2-(Thiophen-2-yl)quinoxaline 97%
2-(Thiophen-2-yl)quinoxaline

Precaución: for Research Use Only

Formula: C₁₂H₈N₂S
MW: 212.27 g/mol
MDL Number: MFCD00067832
CAS Number: 40353-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR