2-Phenylnicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Phenylnicotinic acid, 2-Phenylpyridine-3-carboxylic acid

A451860-250MG A451860-1G
77279-194EA 24.72 USD
77279-194 77616-948
2-Phenylnicotinic acid 97%
2-Phenylnicotinic acid

Precaución: for Research Use Only

Formula: C₁₂H₉NO₂
MW: 199.21 g/mol
MDL Number: MFCD03069417
CAS Number: 33421-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR