2-(2-Methylphenoxy)propanoic acid 95+%

Supplier: Ambeed
Warning

Synonyms: 2-(o-Tolyloxy)propanoic acid

A451803-1G A451803-250MG A451803-5G
77241-042EA 77.25 USD
77241-042 77241-044 77241-046
2-(2-Methylphenoxy)propanoic acid 95+%
2-(2-Methylphenoxy)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.21 g/mol
MDL Number: MFCD01122029
CAS Number: 7345-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR