2,3-Dihydrobenzo[b]furan-5-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydrobenzo[b]furan-5-carboxylicacid

A450071-1G A450071-250MG A450071-5G
77265-100EA 25 USD
77265-100 77265-102 77265-104
2,3-Dihydrobenzo[b]furan-5-carboxylic acid 98%
2,3-Dihydrobenzo[b]furan-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈O₃
MDL Number: MFCD00085012
CAS Number: 76429-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR