(S)-1-{(S)-2-[Di(2-furyl)phosphino]ferrocenyl}ethylbis(2-methylphenyl)phosphine 98%
Supplier: Ambeed
|
Warning
|
A449844-100MG
A449844-250MG
A449844-50MG
A449844-1G
77252-766EA
90.64
USD
77252-766
77252-768
77252-770
77746-922
(S)-1-{(S)-2-[Di(2-furyl)phosphino]ferrocenyl}ethylbis(2-methylphenyl)phosphine 98%
(S)-1-{(S)-2-[Di(2-furyl)phosphino]ferrocenyl}ethylbis(2-methylphenyl)phosphine
Precaución: for Research Use Only
Formula:
C₃₄H₃₂FeO₂P₂ MW: 590.41 g/mol |
MDL Number:
MFCD08561169 CAS Number: 849924-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...