4,4-Methylenebis(2-ethyl-6-methylaniline) 98%

Supplier: Ambeed
Warning

A449540-100G A449540-25G
77291-678EA 18.54 USD
77291-678 77291-680
4,4-Methylenebis(2-ethyl-6-methylaniline) 98%
4,4-Methylenebis(2-ethyl-6-methylaniline)

Precaución: for Research Use Only

Formula: C₁₉H₂₆N₂
MW: 282.42 g/mol
MDL Number: MFCD00071555
CAS Number: 19900-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR