4-Bromo-1-(bromomethyl)-2-methoxybenzene 95%

Supplier: Ambeed
Danger

A449360-10G A449360-1G A449360-250MG A449360-25G A449360-5G
77252-756EA 376.98 USD
77252-756 77252-758 77252-760 77252-762 77252-764
4-Bromo-1-(bromomethyl)-2-methoxybenzene 95%
4-Bromo-1-(bromomethyl)-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂O
MW: 279.95 g/mol
MDL Number: MFCD11036356
CAS Number: 854778-42-0
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR