2,4-Dibromoquinoline 96%

Supplier: Ambeed
Danger

A449214-250MG A449214-1G A449214-5G
77252-754EA 21 USD
77252-754 77572-406 77572-408
2,4-Dibromoquinoline 96%
2,4-Dibromoquinoline

Precaución: for Research Use Only

Formula: C₉H₅Br₂N
MDL Number: MFCD00837761
CAS Number: 20151-40-0
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR