1-Fluoro-4-isopropylbenzene 98%

Supplier: Ambeed
Danger

A449120-1G A449120-25G A449120-5G
77291-672EA 58.71 USD
77291-672 77291-674 77291-676
1-Fluoro-4-isopropylbenzene 98%
1-Fluoro-4-isopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁F
MW: 138.18 g/mol
MDL Number: MFCD00017969
CAS Number: 403-39-4
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR