4-Chloro-2-fluoro-1-nitrobenzene 98%

Supplier: Ambeed
Warning

A448948-10G A448948-25G A448948-500G A448948-5G
77291-664EA 17 USD
77291-664 77291-666 77291-668 77291-670
4-Chloro-2-fluoro-1-nitrobenzene 98%
4-Chloro-2-fluoro-1-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₂
MDL Number: MFCD00042211
CAS Number: 700-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR