2-Chloroethyl methanesulfonate 98%

Supplier: Ambeed
Danger

A448817-1G A448817-5G A448817-25G
77265-096EA 18 USD
77265-096 77265-098 77748-911
2-Chloroethyl methanesulfonate 98%
2-Chloroethyl methanesulfonate

Precaución: for Research Use Only

Formula: C₃H₇ClO₃S
MDL Number: MFCD00007560
CAS Number: 3570-58-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR