4-Chloro-3-iodopyridine hydrochloride 95%

Supplier: Ambeed
Warning

A448636-1G A448636-5G
77252-750EA 8 USD
77252-750 77252-752
4-Chloro-3-iodopyridine hydrochloride 95%
4-Chloro-3-iodopyridine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₄Cl₂IN
MDL Number: MFCD28016601
CAS Number: 1987263-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR