Ir(p-F-ppy)3 98%

Supplier: Ambeed
Warning

A448396-100MG A448396-1G A448396-50MG
77252-744EA 169.95 USD
77252-744 77252-746 77252-748
Ir(p-F-ppy)3 98%
Ir(p-F-ppy)3

Precaución: for Research Use Only

Formula: C₃₃H₂₁F₃IrN₃
MW: 708.75 g/mol
MDL Number: MFCD30830351
CAS Number: 370878-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR