Ethyl 2-bromopyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

A448383-100MG A448383-1G A448383-250MG A448383-5G
77265-082EA 107.12 USD
77265-082 77265-084 77265-086 77265-088
Ethyl 2-bromopyrimidine-5-carboxylate 97%
Ethyl 2-bromopyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MW: 231.04 g/mol
MDL Number: MFCD28746631
CAS Number: 1805568-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR