Methyl 3,5-bis(bromomethyl)benzoate 97%

Supplier: Ambeed
Danger

A448350-1G A448350-250MG
77265-080EA 211.15 USD
77265-080 77748-910
Methyl 3,5-bis(bromomethyl)benzoate 97%
Methyl 3,5-bis(bromomethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀Br₂O₂
MW: 321.99 g/mol
MDL Number: MFCD00182471
CAS Number: 29333-41-3
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR