(5-Methyl-2-nitrophenyl)methanol 98%

Supplier: Ambeed
Warning

A448157-1G A448157-250MG A448157-5G
77252-738EA 24.72 USD
77252-738 77252-740 77252-742
(5-Methyl-2-nitrophenyl)methanol 98%
(5-Methyl-2-nitrophenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.17 g/mol
MDL Number: MFCD00007370
CAS Number: 66424-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR