N-(6-Benzoyl-1H-benzo[d]imidazol-2-yl)-2-(1-(thieno[3,2-d]pyrimidin-4-yl)piperidin-4-yl)thiazole-4-carboxamide 98+%
Supplier: Ambeed
|
Warning
|
A448073-10MG
A448073-1MG
A448073-25MG
A448073-50MG
A448073-5MG
77265-070EA
49
USD
77265-070
77265-072
77265-074
77265-076
77265-078
N-(6-Benzoyl-1H-benzo[d]imidazol-2-yl)-2-(1-(thieno[3,2-d]pyrimidin-4-yl)piperidin-4-yl)thiazole-4-carboxamide 98+%
N-(6-Benzoyl-1H-benzo[d]imidazol-2-yl)-2-(1-(thieno[3,2-d]pyrimidin-4-yl)piperidin-4-yl)thiazole-4-carboxamide
Precaución: for Research Use Only
Formula:
C₂₉H₂₃N₇O₂S₂ |
MDL Number:
MFCD18206785 CAS Number: 913822-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...