2-Propoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

A448013-1G A448013-250MG A448013-25G A448013-5G
77279-172EA 32.96 USD
77279-172 77279-174 77279-176 77279-178
2-Propoxyphenylboronic acid 98%
2-Propoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
MW: 180.0087 g/mol
MDL Number: MFCD03427191
CAS Number: 134896-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR