3-((2-Amino-3-chloropyridin-4-yl)thio)-6-chloropyrazin-2-amine 97%

Supplier: Ambeed
Warning

A447733-100MG A447733-1G A447733-250MG
77265-064EA 73.13 USD
77265-064 77265-066 77265-068
3-((2-Amino-3-chloropyridin-4-yl)thio)-6-chloropyrazin-2-amine 97%
3-((2-Amino-3-chloropyridin-4-yl)thio)-6-chloropyrazin-2-amine

Precaución: for Research Use Only

Formula: C₉H₇Cl₂N₅S
MW: 288.16 g/mol
MDL Number: MFCD30530800
CAS Number: 1801693-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR