1-(p-Toluenesulfonyl)indole-3-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 1-Tosyl-1H-indole-3-carbaldehyde

A447441-100MG A447441-1G A447441-250MG
77279-166EA 14 USD
77279-166 77279-168 77279-170
1-(p-Toluenesulfonyl)indole-3-carbaldehyde 98%
1-(p-Toluenesulphonyl)indole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃NO₂S
MDL Number: MFCD00238944
CAS Number: 50562-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR