(S)-2-((tert-Butoxycarbonyl)amino)hex-5-enoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Boc-2-(3-Butenyl)-Gly-OH

A447337-1G A447337-250MG
77241-022EA 954.81 USD
77241-022 77241-024
(S)-2-((tert-Butoxycarbonyl)amino)hex-5-enoic acid 95%
(S)-2-((tert-Butoxycarbonyl)amino)hex-5-enoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MW: 229.27 g/mol
MDL Number: MFCD03092913
CAS Number: 208522-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR